1-(2-(Dimethylamino)ethyl)-1H-pyrazol-4-amine 97%, Ambeed.Inc

Supplier: Ambeed
Danger

A524850-250MG A524850-1G
77169-546EA 41.2 USD
77169-546 77684-360
1-(2-(Dimethylamino)ethyl)-1H-pyrazol-4-amine 97%, Ambeed.Inc
1-(2-(Dimethylamino)ethyl)-1H-pyrazol-4-amine

Precaución: For research use only.

Formula: C₇H₁₄N₄
MW: 154.21 g/mol
MDL Number: MFCD11128204
CAS Number: 1152939-98-4
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR