3,6-Dibromo-4,5-difluorobenzene-1,2-diamine 95%

Supplier: Ambeed
Warning

A524580-100MG A524580-1G A524580-250MG
77169-540EA 44.29 USD
77169-540 77169-542 77169-544
3,6-Dibromo-4,5-difluorobenzene-1,2-diamine 95%
3,6-Dibromo-4,5-difluorobenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₆H₄Br₂F₂N₂
MW: 301.91 g/mol
MDL Number: MFCD29918985
CAS Number: 1345627-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR