4,4',6,6'-Tetramethyl-2,2'-bipyridine 98%
Supplier: Ambeed
|
Warning
|
A524139-100MG
A524139-1G
A524139-250MG
A524139-5G
77197-272EA
17.51
USD
77197-272
77197-274
77197-276
77747-070
4,4',6,6'-Tetramethyl-2,2'-bipyridine 98%
4,4',6,6'-Tetramethyl-2,2'-bipyridine
Precaución: for Research Use Only
Formula:
C₁₄H₁₆N₂ MW: 212.29 g/mol |
MDL Number:
MFCD11042529 CAS Number: 4444-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...