2-(1H-Benzo[d]imidazol-2-yl)ethanamine 98%

Supplier: Ambeed
Warning

A523874-10G A523874-250MG A523874-25G A523874-5G A523874-1G
77169-532EA 191.58 USD
77169-532 77169-534 77169-536 77169-538 77594-626
2-(1H-Benzo[d]imidazol-2-yl)ethanamine 98%
2-(1H-Benzo[d]imidazol-2-yl)ethanamine

Precaución: for Research Use Only

Formula: C₉H₁₁N₃
MW: 161.2037 g/mol
MDL Number: MFCD00462745
CAS Number: 29518-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR