5-(Trifluoromethyl)benzo[d]thiazole-2(3H)-thione 95%

Supplier: Ambeed
Warning

A522721-100MG A522721-1G A522721-250MG
77169-526EA 21.63 USD
77169-526 77169-528 77169-530
5-(Trifluoromethyl)benzo[d]thiazole-2(3H)-thione 95%
5-(Trifluoromethyl)benzo[d]thiazole-2(3H)-thione

Precaución: for Research Use Only

Formula: C₈H₄F₃NS₂
MW: 235.24 g/mol
MDL Number: MFCD01809822
CAS Number: 23420-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR