Hexylene di(acetate) 98%

Supplier: Ambeed
Warning

Synonyms: 1,6-Diacetoxyhexane, Hexylene glycol diacetate, Hexane-1,6-diyl diacetate, 1,6-Hexanediol diacetate

A522528-100G A522528-25G A522528-500G
77197-266EA 37.08 USD
77197-266 77197-268 77197-270
Hexylene di(acetate) 98%
Hexylene di(acetate)

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
MDL Number: MFCD00191655
CAS Number: 6222-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR