2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(2,2,2-Trifluoroacetyl)-1,2,3,4-tetrahydroisoquinoline-7-sulphonyl chloride
A521315-250MG
A521315-25G
A521315-5G
77197-254EA
20.6
USD
77197-254
77197-256
77197-258
2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride 98%
2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride
Precaución: for Research Use Only
Formula:
C₁₁H₉ClF₃NO₃S MW: 327.71 g/mol |
MDL Number:
MFCD01075651 CAS Number: 74291-57-9 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...