3-(4-Methylpiperazin-1-yl)propylamine 98%

Supplier: Ambeed
Danger

A520422-1G A520422-25G A520422-5G
77197-240EA 15 USD
77197-240 77197-242 77197-244
3-(4-Methylpiperazin-1-yl)propylamine 98%
3-(4-Methylpiperazin-1-yl)propylamine

Precaución: for Research Use Only

Formula: C₈H₁₉N₃
MDL Number: MFCD00014616
CAS Number: 4572-03-6
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR