2,4-Difluoro-5-nitrobenzotrifluoride 97%

Supplier: Ambeed
Warning

A519868-100MG A519868-1G A519868-250MG A519868-5G
77197-232EA 21 USD
77197-232 77197-234 77197-236 77197-238
2,4-Difluoro-5-nitrobenzotrifluoride 97%
2,4-Difluoro-5-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂F₅NO₂
MDL Number: MFCD16659985
CAS Number: 35010-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR