1-(4-Benzoylpiperazin-1-yl)propan-1-one 98%

Supplier: Ambeed
Warning

Synonyms: Sunifiram

A519572-1G A519572-25G A519572-5G A519572-250MG
77169-516EA 6.18 USD
77169-516 77169-518 77169-520 77613-634
1-(4-Benzoylpiperazin-1-yl)propan-1-one 98%
1-(4-Benzoylpiperazin-1-yl)propan-1-one

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₂
MW: 246.30 g/mol
MDL Number: MFCD04112755
CAS Number: 314728-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR