4-Morpholinobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Morpholinobenzoic acid, 4-(4-Morpholinyl)benzoic acid
A518939-100G
A518939-10G
A518939-1G
A518939-250MG
A518939-5G
77169-504EA
455.26
USD
77169-504
77169-506
77169-508
77169-510
77684-348
4-Morpholinobenzoic acid 97%
4-Morpholinobenzoic acid
Precaución: For research use only.
Formula:
C₁₁H₁₃NO₃ MW: 207.2300 g/mol |
MDL Number:
MFCD00588379 CAS Number: 7470-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...