3,4-Dihydro-1H-quinoxalin-2-one 95%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dihydroquinoxalin-2(1H)-one

A518635-1G A518635-250MG
77169-500EA 50.47 USD
77169-500 77169-502
3,4-Dihydro-1H-quinoxalin-2-one 95%
3,4-Dihydro-1H-quinoxalin-2-one

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.17 g/mol
MDL Number: MFCD00204133
CAS Number: 59564-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR