4,5-Dibromo-3-chlorothiophene-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A518128-1G A518128-250MG A518128-5G
77169-494EA 74 USD
77169-494 77169-496 77169-498
4,5-Dibromo-3-chlorothiophene-2-carboxylic acid 95%
4,5-Dibromo-3-chlorothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅HBr₂ClO₂S
MDL Number: MFCD28099449
CAS Number: 503308-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR