2,2'-(5-Bromo-1,3-phenylene)dipyridine 95+%

Supplier: Ambeed
Warning

A517911-100MG A517911-1G A517911-250MG
77169-488EA 59.74 USD
77169-488 77169-490 77169-492
2,2'-(5-Bromo-1,3-phenylene)dipyridine 95+%
2,2'-(5-Bromo-1,3-phenylene)dipyridine

Precaución: for Research Use Only

Formula: C₁₆H₁₁BrN₂
MW: 311.17 g/mol
MDL Number: MFCD29049206
CAS Number: 150239-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR