1,2-Dimethyl-1H-imidazole-5-carbaldehyde 95%
Supplier: Ambeed
|
Warning
|
A517233-100MG
A517233-250MG
77197-214EA
121.54
USD
77197-214
77197-216
1,2-Dimethyl-1H-imidazole-5-carbaldehyde 95%
1,2-Dimethyl-1H-imidazole-5-carbaldehyde
Precaución: for Research Use Only
Formula:
C₆H₈N₂O |
MDL Number:
MFCD06589827 CAS Number: 24134-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...