(E)-Ethyl 3-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acrylate 98+%

Supplier: Ambeed
Warning

A516837-1G A516837-250MG A516837-5G
77169-464EA 195.7 USD
77169-464 77169-466 77169-468
(E)-Ethyl 3-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acrylate 98+%
(E)-Ethyl 3-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₇H₂₃BO₄
MW: 302.17 g/mol
MDL Number: MFCD26127274
CAS Number: 1377024-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR