5-Fluoro-2-hydroxybenzamide 97%

Supplier: Ambeed
Warning

A516333-1G A516333-25G A516333-5G
77197-210EA 12 USD
77197-210 77197-212 77747-061
5-Fluoro-2-hydroxybenzamide 97%
5-Fluoro-2-hydroxybenzamide

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD12913381
CAS Number: 56874-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR