Methyl 3-(3-(difluoromethyl)-1-methyl-1H-pyrazol-4-yl)-3-oxopropanoate 95%

Supplier: Ambeed
Warning

A515869-100MG A515869-250MG
77169-458EA 8 USD
77169-458 77169-460
Methyl 3-(3-(difluoromethyl)-1-methyl-1H-pyrazol-4-yl)-3-oxopropanoate 95%
Methyl 3-(3-(difluoromethyl)-1-methyl-1H-pyrazol-4-yl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₉H₁₀F₂N₂O₃
MDL Number: MFCD28101778
CAS Number: 1806680-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR