tert-Butyl 1H-pyrazol-4-ylcarbamate 98%

Supplier: Ambeed
Warning

A515666-100MG A515666-5G
77169-456EA 31.93 USD
77169-456 77684-339
tert-Butyl 1H-pyrazol-4-ylcarbamate 98%
tert-Butyl 1H-pyrazol-4-ylcarbamate

Precaución: For research use only.

Formula: C₈H₁₃N₃O₂
MW: 183.21 g/mol
MDL Number: MFCD12133944
CAS Number: 130106-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR