Methyl-2-amino-2-(4-chlorophenyl)acetate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-amino-2-(4-chlorophenyl)acetate hydrochloride
A515647-1G
A515647-250MG
A515647-5G
77197-200EA
13
USD
77197-200
77197-202
77197-204
Methyl-2-amino-2-(4-chlorophenyl)acetate hydrochloride 95%
Methyl-2-amino-2-(4-chlorophenyl)acetate hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₁Cl₂NO₂ |
MDL Number:
MFCD00196831 CAS Number: 42718-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...