6-Methoxy-4-methylnicotinonitrile 97%

Supplier: Ambeed
Warning

A515283-10G A515283-250MG A515283-5G A515283-1G
77197-176EA 206 USD
77197-176 77197-178 77197-180 77747-057
6-Methoxy-4-methylnicotinonitrile 97%
6-Methoxy-4-methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.16 g/mol
MDL Number: MFCD18818953
CAS Number: 243469-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR