1-(2-Methoxyethyl)-1H-pyrazol-4-amine 98%

Supplier: Ambeed
Warning

A515126-100MG A515126-250MG A515126-1G
77169-452EA 11 USD
77169-452 77169-454 77893-047
1-(2-Methoxyethyl)-1H-pyrazol-4-amine 98%
1-(2-Methoxyethyl)-1H-pyrazol-4-amine

Precaución: for Research Use Only

Formula: C₆H₁₁N₃O
MDL Number: MFCD09965625
CAS Number: 948570-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR