3-Bromo-6-methoxy-1H-pyrrolo[3,2-b]pyridine 97%

Supplier: Ambeed
Warning

A514988-100MG A514988-250MG
77197-172EA 140.08 USD
77197-172 77197-174
3-Bromo-6-methoxy-1H-pyrrolo[3,2-b]pyridine 97%
3-Bromo-6-methoxy-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.06 g/mol
MDL Number: MFCD12963085
CAS Number: 1190317-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR