Imidazo[1,5-a]pyridine-7-carboxylic acid dihydrochloride 95%

Supplier: Ambeed
Warning

A514449-1G A514449-250MG
77169-446EA 77 USD
77169-446 77169-448
Imidazo[1,5-a]pyridine-7-carboxylic acid dihydrochloride 95%
Imidazo[1,5-a]pyridine-7-carboxylic acid dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₈Cl₂N₂O₂
MDL Number: MFCD28099095
CAS Number: 1616526-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR