tert-Butyl (cis-3-formylcyclobutyl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: rel-tert-butyl N-[(1s,3s)-3-formylcyclobutyl]carbamate

A513650-100MG A513650-250MG
77197-160EA 144 USD
77197-160 77197-162
tert-Butyl (cis-3-formylcyclobutyl)carbamate 98%
tert-Butyl (cis-3-formylcyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
MDL Number: MFCD28403245
CAS Number: 171549-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR