N-(3,4-Dihydroxyphenethyl)acetamide 95%

Supplier: Ambeed
Warning

A513582-100MG A513582-1G A513582-250MG A513582-5G
77197-154EA 75 USD
77197-154 77197-156 77197-158 77747-052
N-(3,4-Dihydroxyphenethyl)acetamide 95%
N-(3,4-Dihydroxyphenethyl)acetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00037174
CAS Number: 2494-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR