ID-8 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Methoxyphenyl)-2-methyl-3-nitro-1H-indol-6-ol

A512614-100MG A512614-10MG A512614-1MG A512614-50MG A512614-5MG
77197-144EA 146.26 USD
77197-144 77197-146 77197-148 77197-150 77197-152
ID-8 98%
ID-8

Precaución: for Research Use Only

Formula: C₁₆H₁₄N₂O₄
MW: 298.30 g/mol
MDL Number: MFCD00593367
CAS Number: 147591-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR