1-Bromo-4-[1-(trifluoromethyl)cycloprop-1-yl]benzene 95%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromophenyl)-1-(trifluoromethyl)cyclopropane, 1-Bromo-4-(1-(trifluoromethyl)cyclopropyl)benzene

A512473-100MG A512473-1G A512473-250MG A512473-5G
77169-426EA 42.23 USD
77169-426 77169-428 77169-430 77684-334
1-Bromo-4-[1-(trifluoromethyl)cycloprop-1-yl]benzene 95%
1-Bromo-4-[1-(trifluoromethyl)cycloprop-1-yl]benzene

Precaución: For research use only.

Formula: C₁₀H₈BrF₃
MW: 265.07 g/mol
MDL Number: MFCD16817615
CAS Number: 1227160-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR