4-Methoxy-N-methyl-2-nitrobenzenamine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Methoxy-N-methyl-2-nitroaniline
A512147-10G
A512147-1G
A512147-250MG
A512147-5G
A512147-25G
77197-132EA
107.12
USD
77197-132
77197-134
77197-136
77197-138
77617-192
4-Methoxy-N-methyl-2-nitrobenzenamine 97%
4-Methoxy-N-methyl-2-nitrobenzenamine
Precaución: for Research Use Only
Formula:
C₈H₁₀N₂O₃ MW: 182.1800 g/mol |
MDL Number:
MFCD00962666 CAS Number: 3360-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...