4-Bromo-2-tert-butyl-aniline 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-(tert-butyl)aniline, 4-Bromo-2-tert-butylaniline

A512133-1G A512133-250MG
77197-128EA 111.24 USD
77197-128 77197-130
4-Bromo-2-tert-butyl-aniline 98%
4-Bromo-2-tert-butyl-aniline

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrN
MW: 228.129 g/mol
MDL Number: MFCD09965991
CAS Number: 850012-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR