2-Oxo-1,2-dihydro-[1,8]naphthyridine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylic acid

A511715-100MG A511715-1G A511715-250MG
77169-420EA 29.87 USD
77169-420 77169-422 77169-424
2-Oxo-1,2-dihydro-[1,8]naphthyridine-3-carboxylic acid 98%
2-Oxo-1,2-dihydro-[1,8]naphthyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MW: 190.156 g/mol
MDL Number: MFCD06659017
CAS Number: 5175-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR