1,3,5-Tri(9H-carbazol-9-yl)benzene 98%

Supplier: Ambeed
Warning

A511075-1G A511075-250MG
77197-122EA 124.63 USD
77197-122 77197-124
1,3,5-Tri(9H-carbazol-9-yl)benzene 98%
1,3,5-Tri(9H-carbazol-9-yl)benzene

Precaución: for Research Use Only

Formula: C₄₂H₂₇N₃
MW: 573.70 g/mol
MDL Number: MFCD03844810
CAS Number: 148044-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR