1-Benzyl-3-methylpiperazine-1,3-dicarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl 3-methyl piperazine-1,3-dicarboxylate

A510376-1G A510376-5G
77197-118EA 54.59 USD
77197-118 77197-120
1-Benzyl-3-methylpiperazine-1,3-dicarboxylate 95%
1-Benzyl-3-methylpiperazine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₄
MW: 278.3037 g/mol
MDL Number: MFCD03001724
CAS Number: 129799-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR