4-Hydroxy-3,5-diisopropylbenzoic acid 95%

Supplier: Ambeed
Warning

A510292-100MG A510292-1G A510292-250MG
77169-414EA 31 USD
77169-414 77169-416 77169-418
4-Hydroxy-3,5-diisopropylbenzoic acid 95%
4-Hydroxy-3,5-diisopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MDL Number: MFCD00490203
CAS Number: 13423-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR