1-(4-Fluorophenyl)-2-thiourea 98%

Supplier: Ambeed
Danger

Synonyms: N-(4-Fluorophenyl)thiourea, 4-Fluorophenylthiourea

A510066-1G A510066-25G A510066-5G
77197-112EA 9.27 USD
77197-112 77197-114 77197-116
1-(4-Fluorophenyl)-2-thiourea 98%
1-(4-Fluorophenyl)-2-thiourea

Precaución: for Research Use Only

Formula: (C₆H₄F)NHC(S)NH₂
MW: 170.21 g/mol
MDL Number: MFCD00041180
CAS Number: 459-05-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR