1-(6-Amino-3,3-dimethylindolin-1-yl)ethanone 98%

Supplier: Ambeed
Warning

A509935-100MG A509935-1G A509935-250MG
77197-096EA 69.01 USD
77197-096 77197-098 77197-100
1-(6-Amino-3,3-dimethylindolin-1-yl)ethanone 98%
1-(6-Amino-3,3-dimethylindolin-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O
MDL Number: MFCD07636699
CAS Number: 453562-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR