6-Bromo-1H-indazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A508476-100MG A508476-10G A508476-1G A508476-250MG A508476-5G
77169-394EA 8 USD
77169-394 77169-396 77169-398 77169-400 77169-402
6-Bromo-1H-indazole-3-carboxylic acid 97%
6-Bromo-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MDL Number: MFCD05663977
CAS Number: 660823-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR