3-Iodo-N-methylpyridin-4-amine 97%

Supplier: Ambeed
Warning

A508425-1G A508425-250MG
77197-086EA 69.01 USD
77197-086 77197-088
3-Iodo-N-methylpyridin-4-amine 97%
3-Iodo-N-methylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₆H₇IN₂
MW: 234.03 g/mol
MDL Number: MFCD18820783
CAS Number: 680859-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR