1,1,3,3-Tetraethoxy-2-methylpropane 95%

Supplier: Ambeed
Warning

A507179-1G A507179-25G
77197-084EA 23 USD
77197-084 77594-592
1,1,3,3-Tetraethoxy-2-methylpropane 95%
1,1,3,3-Tetraethoxy-2-methylpropane

Precaución: for Research Use Only

Formula: C₁₂H₂₆O₄
MDL Number: MFCD00541470
CAS Number: 10602-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR