Chloromethyl methyl carbonate 95%

Supplier: Ambeed
Danger

A505505-10G A505505-1G A505505-25G
77169-378EA 14.42 USD
77169-378 77169-380 77169-382
Chloromethyl methyl carbonate 95%
Chloromethyl methyl carbonate

Precaución: for Research Use Only

Formula: C₃H₅ClO₃
MW: 124.52 g/mol
MDL Number: MFCD19232041
CAS Number: 40510-81-4
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR