6-Fluoro-5-methylnicotinic acid 95%

Supplier: Ambeed
Warning

A505330-250MG A505330-5G
77169-376EA 24 USD
77169-376 77893-015
6-Fluoro-5-methylnicotinic acid 95%
6-Fluoro-5-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD07781235
CAS Number: 885267-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR