Methyl-5-amino-4-methylpyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-amino-4-methylpicolinate

A504936-100MG A504936-1G A504936-250MG
77169-370EA 53.56 USD
77169-370 77169-372 77169-374
Methyl-5-amino-4-methylpyridine-2-carboxylate 97%
Methyl-5-amino-4-methylpyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD18803232
CAS Number: 868551-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR