1,6-Dichloroisoquinoline 97%

Supplier: Ambeed
Warning

A503495-100MG A503495-10G A503495-1G A503495-250MG A503495-25G A503495-5G
77169-354EA 10 USD
77169-354 77169-356 77169-358 77169-360 77893-012 77893-013
1,6-Dichloroisoquinoline 97%
1,6-Dichloroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅Cl₂N
MW: 198.0500 g/mol
MDL Number: MFCD06738660
CAS Number: 630421-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR