1,3-Dimethylimidazolium iodide 97%
Supplier: Ambeed
|
Warning
|
A503375-100G
A503375-10G
A503375-25G
A503375-500G
A503375-5G
77197-052EA
43
USD
77197-052
77197-054
77197-056
77197-058
77197-060
1,3-Dimethylimidazolium iodide 97%
1,3-Dimethylimidazolium iodide
Precaución: for Research Use Only
Formula:
C₅H₉IN₂ |
MDL Number:
MFCD00159685 CAS Number: 4333-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...