4-Phenyl-1,3-thiazole-2-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 4-Phenylthiazole-2-carbaldehyde

A503314-100MG A503314-250MG
77169-350EA 49.44 USD
77169-350 77169-352
4-Phenyl-1,3-thiazole-2-carbaldehyde 95%
4-Phenyl-1,3-thiazole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NOS
MW: 189.234 g/mol
MDL Number: MFCD02665589
CAS Number: 75390-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR