CNX-774 98+%

Supplier: Ambeed
Warning

A503177-100MG A503177-10MG A503177-1MG A503177-2MG A503177-50MG A503177-5MG
77197-032EA 182 USD
77197-032 77197-034 77197-036 77197-038 77197-040 77197-042
CNX-774 98+%
CNX-774

Precaución: for Research Use Only

Formula: C₂₆H₂₂FN₇O₃
MDL Number: MFCD26405992
CAS Number: 1202759-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR