N-Phenyldibenzo[b,d]furan-4-amine 99%

Supplier: Ambeed
Warning

A502928-1G A502928-5G
77169-342EA 21.63 USD
77169-342 77169-344
N-Phenyldibenzo[b,d]furan-4-amine 99%
N-Phenyldibenzo[b,d]furan-4-amine

Precaución: for Research Use Only

Formula: C₁₈H₁₃NO
MW: 259.30 g/mol
CAS Number: 743453-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR