6-Iodoquinolin-4-amine 95+%

Supplier: Ambeed
Warning

A502790-100MG A502790-250MG
77169-338EA 52.53 USD
77169-338 77169-340
6-Iodoquinolin-4-amine 95+%
6-Iodoquinolin-4-amine

Precaución: for Research Use Only

Formula: C₉H₇IN₂
MW: 270.06 g/mol
MDL Number: MFCD06658311
CAS Number: 40107-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR