1,2-Dihexadecanoyl-sn-glycero-3-[phospho-rac-(1-glycerol)]sodiumsalt 97%

Supplier: Ambeed
Warning

Synonyms: rac-DPPG sodium

A502429-250MG A502429-100MG A502429-1G
77197-030EA 66.95 USD
77197-030 77617-146 77747-029
1,2-Dihexadecanoyl-sn-glycero-3-[phospho-rac-(1-glycerol)]sodiumsalt 97%
1,2-Dihexadecanoyl-sn-glycero-3-[phospho-rac-(1-glycerol)]sodiumsalt

Precaución: for Research Use Only

Formula: C₃₈H₇₄NaO₁₀P
MW: 744.95 g/mol
MDL Number: MFCD00070068
CAS Number: 67232-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR