1,3,6-Hexanetricarbonitrile 98%

Supplier: Ambeed
Danger

A502227-1G A502227-25G A502227-5G
77197-024EA 6.18 USD
77197-024 77197-026 77197-028
1,3,6-Hexanetricarbonitrile 98%
1,3,6-Hexanetricarbonitrile

Precaución: for Research Use Only

Formula: C₉H₁₁N₃
MDL Number: MFCD00129792
CAS Number: 1772-25-4
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR